Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ G6PDH (L. mesenteroides), NAD Assayed

Catalog No. 89945333 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2000 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-945-333 2000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-945-333 Supplier Abnova™ Supplier No. P5267
Add to Cart
Add to Cart

Used for Enzyme

#REF!

Specifications

For Use With (Application) Enzyme Activity
Molecular Weight (g/mol) 104kDa
Name G6PDH (L. mesenteroides), NAD Assayed
Preparation Method Native protein chromatographically purified from Leuconostoc mesenteroides
Quantity 2000 U
Storage Requirements Store at 4°C.
Regulatory Status RUO
Common Name G6PDH
Assays Glucose-6-Phosphate Dehydrogenase Assay
Biological Activity ≥360 NAD units per mg protein. One Unit reduces one micromole of NAD per minute at 37°C, pH 7.8, using glucose-6-phosphate as substrate.
Species Bacteria
Protein Tag None
Expression System Native protein chromatographically purified from Leuconostoc mesenteroides
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.