Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ G6PDH (L. mesenteroides), NADP Assayed

Catalog No. 89945335 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-945-335 500 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-945-335 Supplier Abnova™ Supplier No. P5265
Add to Cart
Add to Cart

Used for Enzyme

#REF!

Specifications

For Use With (Application) Enzyme Activity
Formulation In 3.7M ammonium sulfate
Molecular Weight (g/mol) 104kDa
Name G6PDH (L. mesenteroides), NADP Assayed
Preparation Method Native protein chromatographically purified from Leuconostoc mesenteroides
Quantity 500 U
Storage Requirements Store at 4°C. This product is stable for at least 3 months.
Regulatory Status RUO
Common Name G6PDH
Assays Glucose-6-Phosphate Dehydrogenase Assay
Biological Activity ≥200 NADP units per mg protein. One Unit reduces one micromole of NADP per minute at 37°C, pH 7.8, using glucose-6-phosphate as substrate.
Species Bacteria
Protein Tag None
Expression System Native protein chromatographically purified from Leuconostoc mesenteroides
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.