Learn More
Abcam Gabapentin, Alpha2delta Ligand, >99% Purity, Solid, 50 mg
Description
- Molecular Weight: 171.24 Da
- Purity: >99%
- Form: Solid
- Activity: Interacts with the α2δ subunit of L-type voltage-gated Ca2+ channels; disrupts calcium channel trafficking
- Biological Effect: Anticonvulsant and analgesic in vivo
- Storage Buffer: 100% Gabapentin
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Target Molecule | α2δ Subunit (L-type Voltage-Gated Ca²± Channels) |
| Shipping Condition | Ambient |
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Buffer | 100% Gabapentin |
| Formulation | 100% Gabapentin |
| Quantity | 50 mg |
| For Use With (Application) | Anticonvulsant, Analgesic |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.