Learn More
Enzo Life Sciences Gabapentin-lactam (250mg). CAS: 64744-50-9
SDP
Supplier:  Enzo Life Sciences BMLKC1600250
Encompass_Preferred
Gabapentin-lactam but not gabapentin reduces protein aggregates and improves motor performance in a transgenic mouse model of Huntington’s disease. It opens mitochondrial ATP-dependent potassium channels. Purity: ≥95% (GC). Solubility: Soluble in DMSO (40mg/ml) or ethanol (40mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1075
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.