Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gabapentin, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18602883 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-883 500 g Amber Glass Bottle
18-602-881 25 g Amber Glass Bottle
18-602-884 1 kg Amber Glass Bottle
18-602-885 5 kg Poly Pail
18-602-886 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-602-883 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. G1092500GM
Add to Cart
Add to Cart

G1092, 60142-96-3, C9H17NO2

Spectrum™ Chemical Gabapentin, USP is used with other medications to prevent and control seizures and to relieve nerve pain following shingles in adults. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 60142-96-3
Molecular Formula C9H17NO2
Molecular Weight (g/mol) 171.24
InChI Key UGJMXCAKCUNAIE-UHFFFAOYSA-N
IUPAC Name 2-[1-(aminomethyl)cyclohexyl]acetic acid
SMILES NCC1(CC(O)=O)CCCCC1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 98 to 102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.