Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Gabexate mesylate
GREENER_CHOICE

Catalog No. p-4783977
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-981-0 10 mg
17-985-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-981-0 Supplier Tocris Bioscience™ Supplier No. 1798/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Serine protease inhibitor; inhibits thrombin, trypsin, kallikrein and plasmin

Gabexate mesylate is a serine protease inhibitor; Gabexate mesylate inhibits trypsin, plasmin, plasma kallikrein and thrombin (IC50 values are 9.4, 30, 41 and 110 μM respectively). Antithrombotic in vitro and in vivo. Gabexate mesylate also inhibits LPS-induced TNF-α production, probably by inhibiting NF-κB and AP-1 activation. Gabexate mesylate attenuates allergen-induced airway inflammation and eosinophils infiltration in the lung of asthma murine model.

Chemical Identifiers

CAS 56974-61-9
Molecular Formula C17H27N3O7S

Specifications

Quantity 10 mg
Formula Weight 417.48
Purity >98%
Molecular Weight (g/mol) 417.48
Chemical Name or Material Gabexate mesylate
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.