Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GABRA2 Monoclonal Antibody (N399/19), FITC
GREENER_CHOICE

Catalog No. PIMA545726
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545726 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545726 Supplier Invitrogen™ Supplier No. MA545726
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

A 1:100 dilution of MA5-45726 was sufficient for detection of GABA-A R, Alpha2 in 20 μg of mouse brain lysate by ECL immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 55kDa. Does not cross-react with GABA-A Receptor Alpha1. This antibody was formerly sold as clone S399-19.

GABA is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA-A receptors, which are ligand-gated chloride channels. Chloride conductance of these channels can be modulated by agents such as benzodiazepines that bind to the GABA-A receptor. At least 16 distinct subunits of GABA-A receptors have been identified. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABRA2
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N399/19
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4
Gene GABRA2
Gene Accession No. P23576, P26048, P47869
Gene Alias C630048P16Rik; FLJ97076; GABA(A) receptor; GABA(A) receptor subunit alpha-2; GABA(A) receptor, alpha 2; GABAARalpha2; Gabra2; Gabra-2; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, alpha 2; gamma-aminobutyric acid (GABA) A receptor, subunit alpha 2; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 2; gamma-aminobutyric acid A receptor alpha 2; gamma-aminobutyric acid receptor A2 subunit; gamma-aminobutyric acid receptor subunit alpha-2; gamma-aminobutyric acid type A receptor alpha2 subunit
Gene Symbols GABRA2
Host Species Mouse
Immunogen Fusion protein amino acids 350-385 (Cytoplasmic C-terminus) of rat GABA-A Receptor Alpha2.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14395, 2555, 289606
Target Species Human, Mouse, Rat
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.