Learn More
Enzo Life Sciences GAD 65/67 monoclonal antibody (9A6) (100µg)
SDP
Supplier:  Enzo Life Sciences ADIMSA225E
Encompass_Preferred
Clone: 9A6. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human GAD65. UniProt ID: Q05329. Species reactivity: Human, Mouse, Rat. Applications: IHC (PS), WB. Recommended Dilutions: Immunohistochemistry (paraffin sections, 1-10µg/ml) Western Blot (1µg/ml, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol.
Catalog No. 50-200-7352
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.