Learn More
Description
A suggested positive control for immunohistochemical applications is brain. In Western blot, a doublet at ~65/67 kDa has been observed. Rabbit anti Glutamate Decarboxylase 1/2 antibody recognizes both molecular forms of glutamate decarboxylase, GAD1 (GAD67) and GAD2 (GAD65), the enzyme which converts glutamic acid to gamma -aminobutyric acid (GABA), the major inhibitory transmitter in the higher brain region.
Specifications
Specifications
| Antigen | GAD65/GAD67 |
| Applications | ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide |
| Gene | Gad1 |
| Gene Accession No. | P14748, P18088, P48318, P48320, Q05329, Q05683, Q99259 |
| Gene Alias | 6330404F12Rik; 65 kDa glutamic acid decarboxylase; 67 kDa glutamic acid decarboxylase; CPSQ1; EC 4.1.1.15; EP10; FLJ45882; GAD; GAD 1; GAD 2; GAD 65; GAD 67; GAD(65); GAD1; Gad-1; Gad2; Gad-2; GAD25; GAD44; Gad65; GAD-65; GAD67; GAD-67; Glutamate decarboxylase 1; Glutamate decarboxylase 1 (brain); glutamate decarboxylase 1 (brain, 67kDa); glutamate decarboxylase 1 variant GAD67NT; glutamate decarboxylase 2; Glutamate decarboxylase 2 (islet); glutamate decarboxylase 2 (pancreatic islets and brain, 65kDa); glutamate decarboxylase 65 kDa isoform; Glutamate decarboxylase 67 kDa isoform; GLUTAMATE DECARBOXYLASE, 67 KD ISOFORM (GAD-67) (67 KD GLUTAMIC ACID DECARBOXYLASE); Glutamate decarboxylase-2 (pancreas); glutamic acid decarboxylase; glutamic acid decarboxylase 1; glutamic acid decarboxylase 2; glutamic acid decarboxylase 65; SCP |
| Gene Symbols | Gad1, GAD2 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
