Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gadolinium(III) acetate hydrate, 99.9% (REO)

Catalog No. AA8915822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8915822 100 g
AA8915806 5 g
AA8915814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA8915822 Supplier Thermo Scientific Chemicals Supplier No. 08915822
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 100587-93-7 | C6H9GdO6 | 334.38 g/mol

Gadolinium(III) acetate hydrate is used as a laboratory reagent, catalysts, optical glasses, structural ceramics, electrical components and photo-optical material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100587-93-7
Molecular Formula C6H9GdO6
Molecular Weight (g/mol) 334.38
MDL Number MFCD00150116
InChI Key LYQGMALGKYWNIU-UHFFFAOYSA-K
IUPAC Name gadolinium(3+) triacetate
SMILES [Gd+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Boiling Point (decomposition)
Color White
Physical Form Crystals
Quantity 100 g
Odor Odorless
Sensitivity Hygroscopic
Assay Percent Range 99.9% (REO)
Density 1.611
Solubility Information Soluble in water.
Formula Weight 334.39 (Anhydrous)
Chemical Name or Material Gadolinium(III) acetate hydrate
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 240-204-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.