Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gadolinium(III) bromide, ultra dry, 99.99% (metals basis)

Catalog No. AA4367306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4367306 5 g
AA4367314 25 g
AA4367322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4367306 Supplier Thermo Scientific Chemicals Supplier No. 04367306
Add to Cart

CAS: 13818-75-2 | GdBr3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13818-75-2
Molecular Formula GdBr3
MDL Number MFCD00049741

Specifications

Melting Point 770°C
Physical Form Powder
Quantity 5 g
Solubility Information It is soluble in water. Also soluble in THF,1,2-Diethoxyethane,p-dioxane,and alkyl amine.
Formula Weight 396.96
Percent Purity 99.99%
Packaging Ampule
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Gadolinium (III) bromide, Ultra dry
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 237-494-4
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.