Learn More
Gadolinium(III) chloride hexahydrate, REacton™, 99.9% (REO)
Description
Gadolinium(III) chloride hexahydrate is used as a macrophage inhibitor which inhibits the specialized liver macrophages, Kupffer cells that line the walls of the sinusoids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Physical Form | Crystalline Aggregates |
| Quantity | 25 g |
| Sensitivity | Hygroscopic |
| Assay Percent Range | 99.9% (REO) |
| Density | 2.424 g/cm3 |
| Solubility Information | Slightly soluble in water. |
| IUPAC Name | gadolinium(3+);trichloride;hexahydrate |
| Formula Weight | 371.70 (263.61 Anhydrous) |
| Chemical Name or Material | Gadolinium(III) chloride hexahydrate, Rare Earth Oxides Content |
Safety and Handling
| Hazard Category | H315-H319 |
| Hazard Statement | In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. |
| Precautionary Statement | P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362 |
| EINECSNumber | 233-386-6 |
| RTECSNumber | LW4050000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.