Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gadolinium(III) chloride hexahydrate, REacton™, 99.9% (REO)

Catalog No. AA11286A1
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA11286A1 1 kg
AA1128606 5 g
AA1128614 25 g
AA1128622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA11286A1 Supplier Thermo Scientific Chemicals Supplier No. 011286A1
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13450-84-5 | Cl3GdH12O6 | 371.69 g/mol

Gadolinium(III) chloride hexahydrate is used as a macrophage inhibitor which inhibits the specialized liver macrophages, Kupffer cells that line the walls of the sinusoids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13450-84-5
Molecular Formula Cl3GdH12O6
Molecular Weight (g/mol) 371.69
MDL Number MFCD00149732
InChI Key PNYPSKHTTCTAMD-UHFFFAOYSA-K
Synonym gadolinium chloride hexahydrate, gadolinium trichloride hexahydrate, unii-261ex557hi, gadolinium chloride, hydrate 1:6, gadolinium chloride gdcl3 , hexahydrate, gadolinium chloride gdcl3 , hexahydrate 8ci,9ci, gadolinium 3+ trichloride hexahydrate, gadolinium iii chloridehexahydrate, gadolinium chloride, hexahydrate,, gadolinium iii chloride hexahydrate, reacton reo
PubChem CID 197915
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Gd+3]

Specifications

Color White
Physical Form Crystalline Aggregates
Quantity 1 kg
Sensitivity Hygroscopic
Assay Percent Range 99.9% (REO)
Density 2.424
Solubility Information Slightly soluble in water.
IUPAC Name gadolinium(3+) hexahydrate trichloride
Formula Weight 371.70 (263.61 Anhydrous)
Chemical Name or Material Gadolinium(III) chloride hexahydrate, Rare Earth Oxides Content
Hazard Category H315-H319
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 233-386-6
RTECSNumber LW4050000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.