Learn More
Gadolinium(III) Chloride Solution NMR Reference Standard, 0.1 mg/mL in D2O (99.9 atom % D), DSS 0.1%, Water 1%, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Packaging | NMR Tube Size: 3 mm 8 in. |
| Concentration | 0.1 mg/mL in D2O (99.9 atom% D) |
| Linear Formula | GdCl3 |
| MDL Number | MFCD00011024 |
| Quantity | 1 Ea. |
| Formula Weight | 263.61 |
| Grade | NMR Reference Standard |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.