Learn More
Gadolinium(III) oxide, REacton™, 99.99% (REO)
Description
Gadolinium(III) oxide is used as a raw material for various fluorescent compounds, absorption material in atomic reactions, nuclear fuels, magnetic bubble material and screen-sensitivity increasing material. It is also used in glass and electronic industries. It also acts as a neutron shield, a catalyst, a dielectric ceramic and is used for filament coatings, special glasses, lasers, masers and telecommunication. It also serves as a laboratory reagent. It plays an important role as a potential contrast agent for magnetic resonance imaging (MRI).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 2330°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 50 g |
| Assay Percent Range | 99.99% (REO) |
| Solubility Information | Soluble in acid. Insoluble in water. |
| IUPAC Name | gadolinium(3+);oxygen(2-) |
| Formula Weight | 362.5 |
| Density | 7.407 |
| Chemical Name or Material | Gadolinium(III) oxide, Rare Earth Oxides Content |
Safety and Handling
| Hazard Category | H319 |
| Precautionary Statement | P264b-P280i-P305+P351+P338 |
| EINECSNumber | 235-060-9 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.