Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gadolinium(III) oxide, REacton™, 99.99% (REO)

Catalog No. AA1129030
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1129030 250 g
AA1129009 10 g
AA1129018 50 g
AA11290A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA1129030 Supplier Thermo Scientific Chemicals Supplier No. 01129030
Add to Cart

CAS: 12064-62-9 | Gd2O3 | 362.50 g/mol

Gadolinium(III) oxide is used as a raw material for various fluorescent compounds, absorption material in atomic reactions, nuclear fuels, magnetic bubble material and screen-sensitivity increasing material. It is also used in glass and electronic industries. It also acts as a neutron shield, a catalyst, a dielectric ceramic and is used for filament coatings, special glasses, lasers, masers and telecommunication. It also serves as a laboratory reagent. It plays an important role as a potential contrast agent for magnetic resonance imaging (MRI).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12064-62-9
Molecular Formula Gd2O3
Molecular Weight (g/mol) 362.50
MDL Number MFCD00011026
InChI Key CMIHHWBVHJVIGI-UHFFFAOYSA-N
Synonym gadolinia, gadolinium oxide, gadolinium iii oxide, gadolinium trioxide, digadolinium trioxide, gadolinium sesquioxide, gadolinium 3+ oxide, unii-5480d0nhlj, gadolinium 3+ ; oxygen 2-, digadolinium 3+ ion trioxidandiide
PubChem CID 159427
SMILES [O--].[O--].[O--].[Gd+3].[Gd+3]

Specifications

Melting Point 2330°C
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.99% (REO)
Solubility Information Soluble in acid. Insoluble in water.
IUPAC Name gadolinium(3+);oxygen(2-)
Formula Weight 362.5
Density 7.407
Chemical Name or Material Gadolinium(III) oxide, Rare Earth Oxides Content
Hazard Category H319
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 235-060-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.