Learn More
TARGETMOL CHEMICALS INC 8-Chloroadenosine 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T196615MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. 8-Chloroadenosine (NSC-354258) is a 5' AMP-activated protein kinase agonist potentially for the treatment of chronic lymphocytic leukemia. 8-chloroadenosine activity is associated with inhibition of the mTOR pathway.8-Chloroadenosine (NSC-354258) is a nucleoside analog; metabolized in vivo to 8-Chloro-ATP. Incorporates into RNA during transcription and inhibits RNA synthesis. Exhibits cytotoxicity in MM.1S, RPMI-8226, and U266 cancer cell lines; induces G2/M cell cycle arrest and mitotic catastrophe in A549 and H1299 cells. 8-Chloroadenosine (NSC-354258) has been shown to deplete ATP and inhibit tumor growth in hematological malignancies as well as in lung and breast cancer cell lines. Purity 99.48%
Catalog No. 50-287-8411
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.