Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAGE2D Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. p-7204691 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8045 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8045 Supplier Proteintech Group Inc Supplier No. 125321AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GAGE2D, also named as GAGE8 and CT4.8, is expressed in testis and tumor tissues.

Specifications

Antigen GAGE2D
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene GAGE2D
Gene Accession No. Q9UEU5
Gene Alias Cancer/testis antigen 4.8, CT4.8, G antigen 2D, G antigen 8, GAGE 8, GAGE2D, GAGE8
Gene Symbols GAGE2D
Host Species Rabbit
Immunogen GAGE2D Fusion Protein Ag3230
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 729408
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.