Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GAL4 Monoclonal Antibody
GREENER_CHOICE

Catalog No. 338600
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-860-0 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 33-860-0 Supplier Invitrogen™ Supplier No. 338600
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

33-8600 detects the Gal4 DNA Binding Domain by western blot, immunoprecipitation and immunofluorescence corresponding to amino acids 1-147 of the GAL4 DNA binding region.

GAL4 is a transcriptional activator which activates genes necessary for galactose catabolism. The DNA binding domain of GAL4 is contained in the first 74 amino acids and contains a 'zinc finger' sequence motif. However, amino acids 1-147 have been shown to bind to DNA in vitro. While GAL4 (1-74) bind DNA specifically, it is with a lower affinity than the GAL4 (1-147). GAL4 (1-147) binds to a specific 17 base pair nucleotide sequence, but alone is insufficient to activate transcription. Either of the two acidic activating regions, activating region I (amino acids 148-238) or activating region II (amino acids 768-881), must be joined to activate transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAL4
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene GAL4
Gene Accession No. P04386
Gene Alias antigen NY-CO-27; GAL4; Gal-4; GAL81; galactose-responsive transcription factor GAL4; galectin 4; galectin-4; L-36 lactose-binding protein; L36LBP; Lactose-binding lectin 4; lectin, galactoside binding soluble 4; lectin, galactoside-binding, soluble, 4; LGALS4; Regulatory protein GAL4; YPL248C
Gene Symbols GAL4
Host Species Mouse
Immunogen A recombinant protein corresponding to amino acids 1-147 mapping within the amino terminal, DNA binding domain of GAL4.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 855828
Target Species Yeast
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.