Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Galactosidase Goat anti-None, Polyclonal, MP Biomedicals™

Catalog No. MP0856030 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP0856030 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP0856030 Supplier MP Biomedicals, Inc Supplier No. 0856030

Fluorescein-5-isothiocyanate (FITC 'Isomer I') - conjugated goat IgG fraction to β-galactosidase (no cross to E. coli) and buffer salts

Product is fluorescein-5-isothiocyanate (FITC 'Isomer I') - comjugated goat IgG fraction to β-galactosidase (no cross to E. coli) and buffer salts
TRUSTED_SUSTAINABILITY

Specifications

Antigen β-Galactosidase
Applications Immunoassay
Classification Polyclonal
Concentration Total Protein: 29.3 mg/mL F/P Ratio: 5.0 mol/mol Antibody Protein: 3 mg/mL
Conjugate FITC
Description Fluorescein-5-isothiocyanate (FITC 'Isomer I') - comjugated goat IgG fraction to β-galactosidase (no cross to E. coli) and buffer salts
Host Species Goat
Quantity 2 mL
Regulatory Status RUO
Research Discipline Immunoassays
Primary or Secondary Primary
Target Species None
Content And Storage Refrigerated
Form Liquid
Isotype IgG
Reactivity It shows reactivity to β-galactosidase and is tested by Western or dot blot to ensure monospecificity using lysates of D-lac and strain 3.3 of E. coli.
Shipping Condition Cold Pack
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.