Learn More
AdipoGen Galangin (100 mg)
SDP
Supplier:  AdipoGen CDXG0216M100
Encompass_Preferred
Chemical. CAS: 548-83-4. Formula: C15H10O5. MW: 270.24. Galangin is a flavonoid naturally found in herbs used in traditional medicine. It has diverse biological activities, including potent antioxidant, anti-inflammatory, antiobesity, antidiabetic, anti-cancer, neuroprotective, antiviral and antibacterial properties. Galangin has anti-inflammatory actions related to suppression of signaling through NF-kappa/NRLP3 pathway. Galangin induces apoptosis and autophagy in different cancer cells. It is a TRPC5 inhibitor, inhibiting Ca2+ entry into TRPC5 expressing cells. Galangin as a potential DPP-4 inhibitor that improves insulin-stimulated skeletal muscle glucose uptake.
Catalog No. 50-205-4479
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.