Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Galanin (Porcine), Endogenous Galanin Receptor Agonist, 1 mg

Catalog No. 502706831
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6831 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6831 Supplier Abcam Supplier No. AB1411741MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Highly pure, endogenous galanin receptor agonist.

  • Molecular Weight: 3210.5 Da
  • Form: Solid
  • Constituents: 100% Galanin (porcine)
  • Function: Endogenous galanin receptor agonist
  • Storage Conditions: Store at -20°C
  • Shipping Conditions: Ambient - Can Ship with Ice

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Galanin Receptor
Shipping Condition Ambient
Substrate Properties Peptide-based Substrate
Content And Storage Store at -20°C.
Form Solid
Formulation 100% Galanin (porcine)
Quantity 1 mg
For Use With (Application) Galanin receptor agonism
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.