Learn More
Ey Laboratories Inc GALANTHUS NIVALIS GEL-GNA,2ML
Supplier:  Ey Laboratories Inc AK74012
Encompass
GALANTHUS NIVALIS GEL -GNA- IMMOBILIZED LECTIN, 2ML KITGalanthus nivalis Gel -GNA- Immobilized Lectin, 2mL, pre-packed in column with separate elution buffers and instruction booklet.
Catalog No. NC2223459
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.