Learn More
Invitrogen™ Calcitonin/CALCB (Galcanezumab Biosimilar) Recombinant Human Monoclonal Antibody
Shop All Thermo Scientific Products

Description
Endotoxin level <0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt. This product is a Humanized mouse antibody.
Specifications
Specifications
| Antigen | Calcitonin/CALCB (Galcanezumab Biosimilar) |
| Applications | ELISA, SDS-Page |
| Classification | Recombinant Monoclonal |
| Concentration | 4.03 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Calca |
| Gene Accession No. | P01258, P06881 |
| Gene Alias | alpha-type CGRP; CA; Cal1; CAL6; CALC; Calc1; Calca; calcitonin; calcitonin 1; Calcitonin carboxyl-terminal peptide; Calcitonin gene-related peptide 1; calcitonin gene-related peptide I; calcitonin related polypeptide alpha; calcitonin/calcitonin-related polypeptide, alpha; calcitonin; calcitonin gene-related peptide 1; calcitonin-related polypeptide alpha; CCALCI; CCP; Cgrp; CGRP1; CGRP-1; CGRP-I; Ct; Ctn; Katacalcin; KC; MGC126648; PCT; PDN-21; prepro-alpha-calcitonin gene-related peptide; RAT calcitonin; RATCAL6 |
| Gene Symbols | Calca |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.