Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galectin 13 Antibody [PP13/1165], ProSci
SDP

Catalog No. 16819532
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-819-532 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-819-532 Supplier ProSci LLC Supplier No. 337020100
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Galectin 13 Monoclonal Antibody specifically detects Galectin 13 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen Galectin 13
Applications Western Blot, Flow Cytometry, Immunofluorescence, ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PP13/1165
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution IF: 1-2 ug/mL, Flow Cytometry: 0.5-1 ug/million cells in 0.1ml, ELISA: order the BSA-free formulation for coating, IHC (FFPE): 0.5-1 ug/mL for 30 min at RT, Titering of the LGALS13 antibody may be required for optimal performance.
Formulation PBS with 0.1 mg/mL BSA and 0.05% sodium azide
Host Species Mouse
Immunogen A human partial recombinant protein corresponding to amino acids 23-134 was used as the immunogen for the LGALS13 antibody.
Purification Method Protein G affinity chromatography
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29124
Target Species Human
Content And Storage Aliquot and Store at 2-8°C. Avoid freez-thaw cycles.
Form Liquid
Isotype IgG2b κ
Research Category Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.