Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Galectin 9 Monoclonal Antibody (766428)
GREENER_CHOICE

Catalog No. PIMA524369 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524369 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524369 Supplier Invitrogen™ Supplier No. MA524369
  • $594.00
    $489.65 / Each of 1

    Save $104.35

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The protein encoded by this gene is an S-type lectin. It is overexpressed in Hodgkin's disease tissue and might participate in the interaction between the H and RS cells with their surrounding cells and might thus play a role in the pathogenesis of this disease and/or its associated immunodeficiency. Multiple alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Galectin 9
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 766428
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Lgals9
Gene Accession No. O08573
Gene Alias 36 kDa beta-galactoside-binding lectin; AA407335; AI194909; AI265545; ecalectin; Gal9; gal-9; galectin 9; Galectin9; galectin-9; HOM HD 21; HUAT; Lectin galactose binding soluble 9 (Galectin-9); lectin, galactose binding, soluble 5; lectin, galactose binding, soluble 9; Lectin, galactose binding, soluble 9 (Galectin-9); lectin, galactoside binding soluble 9; lectin, galactoside-binding, soluble, 9; LGALS35; Lgals5; LGALS9; LGALS9A; tumor antigen HOM-HD-21; UAT; urate transporter/channel; urate transporter/channel protein
Gene Symbols Lgals9
Host Species Rat
Immunogen E. coli-derived recombinant mouse Galectin-9 (short isoform) Ala2-Thr322.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16859
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.