Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Galectin 9 Monoclonal Antibody (9M1-3), Functional Grade, eBioscience™
GREENER_CHOICE

Catalog No. 16911685
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-911-685 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-911-685 Supplier Invitrogen™ Supplier No. 16911685
  • $734.00
    $550.65 / Each of 1

    Save $183.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

This 9M1-3 monoclonal antibody reacts with human Galectin-9. Galectin-9 is one of fifteen members of the galectin family of glycan-binding proteins. It is expressed on activated T helper cells and Foxp3+ T regulatory cells and plays a critical role in the regulation of immune function. Interaction of Galectin-9 with its ligand, TIM3, induces apoptosis in Th1 and Th17, inhibits Th17 polarization and IFN gamma expression, and drives Treg expansion. Galectin-9 is also thought to contribute to the survival and expansion of some tumors by inhibiting the anti-tumor response and promoting immune evasion. Along with Galectin-1 and Galectin-3, high levels of Galectin-9 expression are associated with poor cancer prognosis, making these potential therapeutic targets. Applications Reported: This 9M1-3 antibody has been reported for use in flow cytometric analysis and functional assays. (Fluorochrome-conjugated 9M1-3 is recommended for use in intracellular flow cytometry). Applications Tested: This 9M1-3 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and h...

The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The protein encoded by this gene is an S-type lectin. It is overexpressed in Hodgkin's disease tissue and might participate in the interaction between the H and RS cells with their surrounding cells and might thus play a role in the pathogenesis of this disease and/or its associated immunodeficiency. Multiple alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Galectin 9
Applications Flow Cytometry, Functional Assay
Classification Monoclonal
Clone 9M1-3
Concentration 1 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2
Gene Lgals9
Gene Accession No. O00182
Gene Alias 36 kDa beta-galactoside-binding lectin; AA407335; AI194909; AI265545; ecalectin; Gal9; gal-9; galectin 9; Galectin9; galectin-9; HOM HD 21; HUAT; Lectin galactose binding soluble 9 (Galectin-9); lectin, galactose binding, soluble 5; lectin, galactose binding, soluble 9; Lectin, galactose binding, soluble 9 (Galectin-9); lectin, galactoside binding soluble 9; lectin, galactoside-binding, soluble, 9; LGALS35; Lgals5; LGALS9; LGALS9A; tumor antigen HOM-HD-21; UAT; urate transporter/channel; urate transporter/channel protein
Gene Symbols Lgals9
Host Species Mouse
Purification Method Affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3965
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.