Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GALK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590338
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590338 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590338 Supplier Invitrogen™ Supplier No. PA590338
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: U-251MG, HepG2, HeLa, MCF7, Mouse heart, Mouse brain, Mouse lung.

Galactokinase is a major enzyme for the metabolism of galactose and its deficiency causes congenital cataracts in the adult population. GALK1 sequence shares the greatest level of conservation, 44. 5% identity with that from E. coli and 34. 6% amino acid identity with the product of the human GALK2 gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GALK1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GALK1
Gene Accession No. P51570, Q9R0N0
Gene Alias AA409894; epididymis secretory protein Li 19; galactokinase; galactokinase 1; galactose kinase; GALK; Galk1; Gk; GK1; Glk; Glk1; HEL-S-19
Gene Symbols GALK1
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14635, 2584
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.