Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gallic acid ethyl ester, 99%

Catalog No. p-3776537
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC279931000 100 g Glass bottle
AC279935000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC279931000 Supplier Thermo Scientific Chemicals Supplier No. 279931000
Only null left
Add to Cart

CAS: 831-61-8 | C9H10O5 | 198.174 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 831-61-8
Molecular Formula C9H10O5
Molecular Weight (g/mol) 198.174
MDL Number MFCD00016430
InChI Key VFPFQHQNJCMNBZ-UHFFFAOYSA-N
Synonym ethyl gallate, gallic acid ethyl ester, phyllemblin, nipagallin a, progallin a, ethylgallate, nipa no. 48, gallic acid, ethyl ester, benzoic acid, 3,4,5-trihydroxy-, ethyl ester, nipa 48
PubChem CID 13250
ChEBI CHEBI:87247
IUPAC Name ethyl 3,4,5-trihydroxybenzoate
SMILES CCOC(=O)C1=CC(=C(C(=C1)O)O)O

Specifications

Melting Point 151°C to 154°C
Color White to Beige
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 100 g
Beilstein 10, 484
Formula Weight 198.18
Percent Purity ≥98.5%
Physical Form Fine Powder
Chemical Name or Material Gallic acid ethyleester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.