Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gallium(III) bromide, ultra dry, 99.998% (metals basis)

Catalog No. AA3569703
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3569703 1 g
AA3569706 5 g
AA3569714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3569703 Supplier Thermo Scientific Chemicals Supplier No. 03569703
Add to Cart

CAS: 13450-88-9 | Br3Ga | 309.435 g/mol

Gallium bromide is used as a catalyst in the alkylation of aromatic compounds by alkyl bromides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13450-88-9
Molecular Formula Br3Ga
Molecular Weight (g/mol) 309.435
MDL Number MFCD00016100
InChI Key SRVXDMYFQIODQI-UHFFFAOYSA-K
Synonym gallium tribromide, gallium bromide gabr3, gallium iii bromide, gallium bromide, gabr3, gallium bromide, gabr3, gallium iii bromide, anhydrous, gallium iii bromide, ultra dry, gallium iii bromide, anhydrous, granular
PubChem CID 83477
IUPAC Name tribromogallane
SMILES [Ga](Br)(Br)Br

Specifications

Boiling Point 279°C
Melting Point 122°C
Color White
Physical Form Crystalline
Quantity 1 g
UN Number UN3260
Solubility Information Soluble in water.
Formula Weight 309.45
Percent Purity 99.998%
Odor Odorless
Packaging Ampule
Sensitivity Moisture sensitive
Density 3.69 g/mL
Assay (metals basis)
Chemical Name or Material Gallium(III) bromide, Ultra dry
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 236-609-5
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.