Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gallium(III) fluoride, 99.995% (metals basis)

Catalog No. p-7049317
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1398703 1 g
AA1398706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1398703 Supplier Thermo Scientific Chemicals Supplier No. 01398703
Only null left
Add to Cart

CAS: 7783-51-9

Gallium(III) fluoride is employed in the preparation and study of optical fluorogallate glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-51-9
MDL Number MFCD00011021

Specifications

Boiling Point ∼1000°C
Melting Point 800°C (sublimation)
Color White
Physical Form Powder
Quantity 1 g
UN Number UN3260
Merck Index 14,4354
Solubility Information Soluble in water (0.02g/L at 200C).
Formula Weight 126.72
Percent Purity 99.999%
Odor Odorless
Sensitivity Moisture sensitive
Density 4.47 g/mL
Assay (metals basis)
Chemical Name or Material Gallium(III) fluoride
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 232-004-5
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.