Learn More
Gallium(III) sulfide, 99.999% (metals basis), Thermo Scientific Chemicals
Description
Gallium(III) sulfide is used in semiconductor materials, functional ceramic materials, fluorescent powder and photoelectric materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1255°C |
| Physical Form | Powder |
| Quantity | 5 g |
| UN Number | UN3260 |
| Solubility Information | It dissolves in water slowly,which is easy to dissolve in concentrated alkaline to get gallate salts. It is also dissolves in HCL and nitric acid. |
| Formula Weight | 235.63 |
| Percent Purity | 99.999% |
| Odor | Odorless |
| Sensitivity | Moisture sensitive |
| Density | 3.77 g/mL |
| Show More |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S. |
| EINECSNumber | 234-692-2 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.