Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gallium Indium Tin eutectic, 99.99% (metals basis)

Catalog No. AA1463409
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1463409 10 g
AA1463403 1 g
AA1463418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1463409 Supplier Thermo Scientific Chemicals Supplier No. 01463409
Add to Cart

Ga:In:Sn; 62:22:16 wt.%

Gallium-Indium eutectic (EGAIn) is an electrically conductive fluid metal. It useful as a contact electrode for the electrical characterization of thin film organic and semiconductor devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gallium Indium Tin eutectic
Melting Point 10.7°C
Color White
Physical Form Liquid
Quantity 10 g
Assay Percent Range 99.99% (Metals basis)
Molecular Formula Ga:In:Sn; 62:22:16 wt.%
MDL Number MFCD00144388
Solubility Information Insoluble in water or organic solvents.
Odor Odorless
Density 6.359
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.