Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gallium phosphide, 99.999% (metals basis)

Catalog No. AA1226609
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1226609 10 g
AA1226604 2 g
AA1226618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1226609 Supplier Thermo Scientific Chemicals Supplier No. 01226609
Add to Cart

For Use in Semiconducting Devices | CAS: 12063-98-8 | GaP | 100.70 g/mol

Gallium phosphide (GaP) is used in semiconductor devices, and in the manufacture of light-emitting diodes (LEDs) doped with other elements or in combination with gallium arsenide phosphide. With appropriate dopants, p-type and n-type semiconductors can be produced. It is used in optical systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12063-98-8
Molecular Formula GaP
Molecular Weight (g/mol) 100.70
MDL Number MFCD00016109
InChI Key HZXMRANICFIONG-UHFFFAOYSA-N
Synonym gallium phosphide, gallium monophosphide, unii-3j421f73dv, ccris 4019, gallium phosphide, polycryst, gallium phosphide trace metals basis, gallium phosphide trace metals basis 1g, gallium phosphide, single crystal substrate , <111>, diam. x thickness 2 in. x 0.5 mm
PubChem CID 82901
IUPAC Name gallanylidynephosphane
SMILES P#[Ga]

Specifications

Melting Point 1465°C
Color Orange
Physical Form Pieces
Quantity 10 g
Assay Percent Range 99.999% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 100.69
Density 4.13 g/cm3
Chemical Name or Material Gallium phosphide
Hazard Category H319-H335
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
EINECSNumber 235-057-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.