Learn More
TARGETMOL CHEMICALS INC G150 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T11344
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. G150 inhibited interferon expression triggered by DSDNA with IC50 of 10.2 nM.G150 is an effective and highly selective inhibitor of human cyclic GMP-AMP synthase (H-CGAS). purity: 97%
Catalog No. 50-258-1539
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.