Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GALNS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522098 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522098 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522098 Supplier Invitrogen™ Supplier No. PA522098
  • $661.00
    $541.65 / Each of 1

    Save $119.35

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Recommended positive controls: H1299, Neuro2A, C2C12, PC-12, Rat2. Predicted reactivity: Mouse (89%), Rat (90%), Zebrafish (81%), Xenopus laevis (84%), Xenopus Tropicalis (80%), Dog (92%), Pig (90%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes N-acetylgalactosamine-6-sulfatase whichis a lysosomal exohydrolase required for the degradation of theglycosaminoglycans, keratan sulfate, and chondroitin 6-sulfate.Sequence alterations including point, missense and nonsensemutations, as well as those that affect splicing, result in adeficiency of this enzyme. Deficiencies of this enzyme lead toMorquio A syndrome, a lysosomal storage disorder.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GALNS
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene GALNS
Gene Accession No. P34059, Q32KJ6, Q571E4
Gene Alias Chondroitinase; chondroitinsulfatase; galactosamine (N-acetyl)-6-sulfatase; galactosamine (N-acetyl)-6-sulfate sulfatase; galactose-6-sulfate sulfatase; GalN6S; GALNAC6S; GalNAc6S sulfatase; Galns; GAS; mFLJ00319; MPS4A; N-acetylgalactosamine-6-sulfatase; N-acetylgalactosamine-6-sulfate sulfatase
Gene Symbols GALNS
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GALNS. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2588, 292073, 50917
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.