Learn More
United Scientific™ Galvanometer, DC
SDPSCI_ED
Catalog No. S15990 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Amperage:
±50-0-50 μA
±500-0-500 μA
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Amperage
S15990 ±500-0-500 μA
S15730 ±50-0-50 μA
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S15990 Supplier United Scientific™ Supplier No. MGV002
Add to Cart
Add to Cart
Ideal option for classroom electricity demonstrations.
  • DC galvanometer mounted in a black plastic case.
  • Scale length of 75 mm, with ± 2.5% accuracy
  • Features 5-way binding post and an external zero-setting screw
  • Not protected against prolonged or significant overload
  • Stackable for convenient storage
Specifications
For Use With (Application) Classroom Demonstrations
Display Analog
Quantity 1 Ea.
Product Type Galvanometer, DC
Amperage ±500-0-500 μA
Warning: Cancer and reproductive harm