Learn More
Selleck Chemical LLC Gambogic Acid 2752-65-0 Guttatic Acid, Guttic Acid
SDP
Supplier:  Selleck Chemical LLC S24485MG
Encompass_Preferred
Gambogic Acid (Guttatic, Guttic) activates caspases with EC50 of 0.78-1.64 μM and competitively inhibits Bcl-XL, Bcl-2, Bcl-W, Bcl-B, Bfl-1 and Mcl-1 with IC50 of 1.47, 1.21, 2.02, 0.66, 1.06 and 0.79 μM, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1911067