Learn More
Enzo Life Sciences Gambogic amide (1mg). CAS: 286935-60-2
SDP
Supplier:  Enzo Life Sciences BMLN1590001
Encompass_Preferred
A novel small molecule NGF mimic discovered using a cell-based chemical genetic screen for TrkA agonists. Gambogic amide selectively binds to TrkA, triggers its tyrosine phosphorylation, elicits PI3-kinase/Akt and MAPK activation, provokes neurite outgrowth in PC12 cells and prevents neuronal cell death. EC50 for antiapoptotic activity is 5nM (see reference S.-W. Jang, et al. 2007). Purity: ≥95% (HPLC). Solubility: Soluble in DMSO (20mg/ml) or ethanol (20mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1174
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.