Learn More
Sigma Aldrich Fine Chemicals Biosciences Gamborg's B-5 Basal Salt Mixture | 500 GR
Supplier:  Sigma Aldrich Fine Chemicals Biosciences G57681L
Encompass
Gamborg's B-5 Basal Salt Mixture | Purity: N/A | Mol Wt: N/A | CAS No: N/A | MFCD: N/A | 500gA powdered basal salt formulation for preparing Gamborg's B-5 Basal Medium, a plant tissue culture medium. Provides essential macro- and micronutrients as well as vitamins to support proliferation and growth of plant cells and tissues in vitro. Widely used in plant biology research and biotechnology applications involving plant tissue culture.
Catalog No. NC2403106
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.