Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

γ-BHC (Lindane) Standard, 100 μg/mL, Methanol, 1 mL

Catalog No. USPP1401
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-PP-1401 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-PP-1401 Supplier Agilent Technologies Supplier No. PP1401
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 58-89-9 , 67-56-1
Molecular Formula C6H6Cl6
Molecular Weight (g/mol) 290.81
MDL Number MFCD00135947
InChI Key JLYXXMFPNIAWKQ-UHFFFAOYSA-N
Synonym gamma-HCH, Lindane
PubChem CID 727
ChEBI CHEBI:24536
IUPAC Name 1,2,3,4,5,6-hexachlorocyclohexane
SMILES ClC1C(Cl)C(Cl)C(Cl)C(Cl)C1Cl

Specifications

CAS Max % 99.99, 0.01
Analyte or Component Names Methanol,gamma-BHC
Element Methanol,gamma-BHC
Solvent or Matrix Methanol
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material gamma-BHC Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.