Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ gamma Catenin Monoclonal Antibody (PG-11E4)
GREENER_CHOICE

Catalog No. 138500
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-850-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-850-0 Supplier Invitrogen™ Supplier No. 138500
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

gamma Catenin Monoclonal Antibody for ICC/IF, IHC, IP, Western Blot

JUP is a major cytoplasmic protein which is the only known constituent common to sub membranous plaques of both desmosomes and intermediate junctions. This protein forms distinct complexes with cadherins and desmosomal cadherins and is a member of the catenin family since it contains a distinct repeating amino acid motif called the armadillo repeat.
TRUSTED_SUSTAINABILITY

Specifications

Antigen gamma Catenin
Applications Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone PG-11E4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene JUP
Gene Accession No. P14923, Q02257
Gene Alias ARVD12; catenin (cadherin-associated protein), gamma 80kDa; Catenin gamma; cb80; cell-adhesion protein plakoglobin; Ctnng; D930025P04Rik; danPG; Desmoplakin III; desmoplakin-3; desmosomal protein; DP3; DPIII; fb11b06; gama-catenin; gamma-catenin; gamma-catenin (plakoglobin); junction plakoglobin; junction plakoglobin a; Jup; jup protein; jupa; OTTHUMP00000164735; PDGB; PG; PKGB; plakoglobin; wu:fb11b06
Gene Symbols JUP
Host Species Mouse
Immunogen Fusion protein consisting of the maltose binding protein fused to full length human plakoglobin protein.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16480, 3728
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.