Learn More
Abcam Gamma H2A.X Staining Kit, 20 Test
Description
- Stains H2A.X
- Suitable for fluorescence microscopy
- Kit designed for detection
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| No. of Reactions | 20 Tests |
| Target Molecule | Gamma H2A.X |
| Format | Kit |
| Detection Method | Fluorescence |
| Product Type | Protein Stain Kit |
| Quantity | 20 Tests |
| For Use With (Application) | Fluorescence microscopy |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.