Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals gamma-Secretase Inhibitor I

Catalog No. AAJ64068LB0
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64068LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64068LB0 Supplier Thermo Scientific Chemicals Supplier No. J64068LB0
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 133407-83-7 | C26H41N3O5, C26H41N3O5 | 475.63 g/mol

Cell permeable. Inhibits --secretase, a protease that cleaves the amyloid precursor protein (APP) to generate --amyloid protein

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133407-83-7
Molecular Formula C26H41N3O5
Synonym Z-Leu-Leu-Nle-CHO (Nle = Norleucine);

Specifications

Quantity 5 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) Cell permeable. Inhibits →-secretase, a protease that cleaves the amyloid precursor protein (APP) to generate →-amyloid protein
Formula Weight 475.62
Sensitivity Hygroscopic
Product Type Gamma-Secretase Inhibitor I
Form Solid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.