Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ gamma Synuclein Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01670515 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-515 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-515 Supplier Invitrogen™ Supplier No. PA118262
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Reconstitute in 20 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Glycerol (1:1) may be added for an additional stability. Avoid repetitive freeze/thaw cycles.

This gene encodes a member of the synuclein family of proteins which are believed to be involved in the pathogenesis of neurodegenerative diseases. Mutations in this gene have also been associated with breast tumor development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen gamma Synuclein
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SNCG
Gene Accession No. O76070, Q63544
Gene Alias BCSG1; Breast cancer-specific gene 1 protein; C79089; Gamma-synuclein; persyn; PRSN; Sensory neuron synuclein; SNCG; SR; Synoretin; synuclein gamma; synuclein, gamma; synuclein, gamma (breast cancer-specific protein 1)
Gene Symbols SNCG
Host Species Sheep
Immunogen Synthetic peptide corresponding to residues E(114) K E E V A E E A Q S G G D(127) of human gamma synuclein protein.
Purification Method Affinity chromatography
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64347, 6623
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.