Learn More
Abcam Gamma-Tocotrienol, Form Of Vitamin E, 1 mg
Description
- Molecular Weight: 410.6 Da
- Purity: >98%
- Form: Liquid
- Storage Buffer: 99% Ethanol, 1% γ-Tocotrienol
- Key Properties: Form of vitamin E, antioxidant agent
- Biological Activity: Induces apoptosis in cancer cells, shows antitumor and antihypertensive effects in vivo, orally active
- Storage Temperature: Store at -20°C
- Shipping Temperature: Ambient - Can Ship with Ice
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Vitamin E (γ-Tocotrienol) |
| Shipping Condition | Ambient |
| Product Type | Antioxidant |
| Buffer | 99% ethanol, 1% γ-tocotrienol |
| Content And Storage | Store at -20°C. |
| Form | Liquid |
| Formulation | 99% Ethanol, 1% γ-Tocotrienol |
| Quantity | 1 mg |
| For Use With (Application) | Antioxidant agent, Cancer research-apoptosis |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.