Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Gamma-Tocotrienol, Vitamin E Form, Liquid, 10 mg

Catalog No. 502705716
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5716 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5716 Supplier Abcam Supplier No. AB14444810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antioxidant form of vitamin E that induces apoptosis in cancer cells.

  • Molecular Weight: 410.6 Da
  • Purity: >98%
  • Form: Liquid
  • Properties: Antioxidant agent, induces apoptosis in cancer cells, shows antitumor and antihypertensive effects in vivo, and is orally active
  • Storage: Store at -20°C
  • Shipping: Ambient - Can Ship with Ice
  • Composition: 99% Ethanol, 1% γ-Tocotrienol

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Vitamin E (γ-Tocotrienol)
Substrate Properties Chemical Substrate
Product Type Antioxidant
Shipping Condition Ambient
Buffer 99% ethanol, 1% γ-tocotrienol
Content And Storage Store at -20°C.
Form Liquid
Formulation 99% Ethanol, 1% γ-Tocotrienol
Quantity 10 mg
For Use With (Application) Antioxidant agent, Cancer research-apoptosis induction, Antitumor and antihypertensive research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.