Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ gamma Tubulin Monoclonal Antibody (4D11), Alexa Fluor™ 647
GREENER_CHOICE

Catalog No. PIMA1850A64 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA1850A64 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA1850A64 Supplier Invitrogen™ Supplier No. MA1850A647
  • $538.18
    $440.65 / Each of 1

    Save $97.53

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

The MA1-850-A647 immunogen is a recombinant protein containing residues A A T R P D Y I S W G T Q E Q of human gamma Tubulin.

This gene encodes a protein that interacts with thyroid hormone receptors, also known as hormone-dependent transcription factors. The gene product interacts specifically with the ligand binding domain. This gene is one of several that may play a role in early-stage non-small cell lung cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen gamma Tubulin
Applications Immunocytochemistry
Classification Monoclonal
Clone 4D11
Concentration 1.0 mg/mL
Conjugate Alexa Fluor 647
Formulation PBS with 1.0mg/mL BSA and 0.05% sodium azide
Gene TUBG1
Gene Accession No. P23258, P83887, Q8VCK3, Q9NRH3
Gene Alias 1500010O08Rik; AI451582; AI503389; AI504772; CDCBM4; centrosome marker; gamma tubulin; gamma-1-tubulin; Gamma-2-tubulin; gamma-tubulin; gamma-tubulin complex component 1; GCP-1; similar to tubulin, gamma 1; Tubg; TUBG1; tubg1.L; TUBG2; TUBGCP1; Tubgl; tubulin gamma 1; tubulin gamma 1 L homeolog; tubulin gamma 2; Tubulin gamma-1 chain; Tubulin gamma-2 chain; tubulin, gamma 1; tubulin, gamma 2; tubulin, gamma polypeptide; Unknown (protein for MGC:133882); XELAEV_18002398mg; xgam; zgc:55894
Gene Symbols TUBG1, Tubg2
Host Species Mouse
Immunogen Recombinant protein containing residues A A T R P D Y I S W G T Q E Q of human gamma Tubulin.
Purification Method Protein G
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103733, 103768, 27175, 7283
Target Species Human, Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.