Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gamma tubulin Rabbit anti-Canine, Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501735251 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5251 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5251 Supplier Proteintech Group Inc Supplier No. 261951AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody can label the centrosome structure.

This gene encodes a protein that interacts with thyroid hormone receptors, also known as hormone-dependent transcription factors. The gene product interacts specifically with the ligand binding domain. This gene is one of several that may play a role in early-stage non-small cell lung cancer.

Specifications

Antigen gamma tubulin
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TUBG1
Gene Accession No. P23258, P83887, Q9GKK5
Gene Alias Gamma 1 tubulin, GCP 1, TUBG, TUBG1, TUBGCP1, Tubulin gamma 1 chain, tubulin, gamma 1, tubulin-gamma
Gene Symbols TUBG1
Host Species Rabbit
Immunogen gamma tubulin Fusion Protein Ag23575
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103733, 403695, 7283
Target Species Canine, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.