Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva GammaBind™ Plus Sepharose™ Medium
GSA_VA

Catalog No. 45500106 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-500-106 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-500-106 Supplier Cytiva Supplier No. 17088604
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Engineered for better binding to rat and mouse IgG subclasses

  • Recombinant Protein G type 3 coupled to Sepharose CL-6B for easy binding and fast separation
  • More enhanced binding capabilities than GammaBind G at both laboratory and process scale
  • Yields up to 18mg/mL mouse IgG
  • Binds to all IgG subclasses from human, mouse, and rat; binds total IgG from guinea pig, rabbit, goat, cow, sheep, and horse

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type GammaBind Plus Sepharose
Ligand Recombinant GammaBind G type 3 lacking albumin-binding region
Particle Size 90 μm
Content And Storage 2°C to 8°C, 20% Ethanol
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.